Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claudin 11/OSPOligodendrocyte Marker Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984564
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4564 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4564 Supplier Bioss Supplier No. BS2183R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The claudin superfamily consists of many structurally related proteins in humans. These proteins, which include claudin 1 through 18, are important structural and functional components of tight junctions in paracellular transport. Claudins are located in both epithelial and endothelial cells in all tight junction-bearing tissues. Claudins, which consist of four trans-membrane domains and two extracellular loops, make up tight junction strands. Claudin-11 is an oligodendrocyte-specific protein that is expressed in the tight junctions of Sertoli cells and myelin sheaths in mice.

Specifications

Antigen Claudin 11/OSPOligodendrocyte Marker
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Cldn11
Gene Accession No. O75508
Gene Alias BOS_1062; claudin 11; claudin 11 (oligodendrocyte transmembrane protein); claudin 11-like; Claudin11; Claudin-11; CLDN11; LOC100352273; Oligodendrocyte transmembrane protein; Oligodendrocyte-specific protein; OSP; Otm
Gene Symbols Cldn11
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Claudin-11.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5010
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.