Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claudin-12 Antibody (393314), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4284K
Description
Claudin-12 Monoclonal antibody specifically detects Claudin-12 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
Claudin-12 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
9069 | |
Store at 4°C in the dark. | |
IgG2a |
Flow Cytometry, Immunocytochemistry, CyTOF | |
393314 | |
Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
claudin 12, claudin-12 | |
HEK293 human embryonic kidney cell line transfected with human Claudin-12, Met1-Thr244, Accession # NP_036261 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction