Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claudin 12 Polyclonal Antibody, Abbomax

Catalog No. 502203533
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3533 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3533 Supplier Abbomax Inc Supplier No. 50010724
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: MCF-7 Cellular location: Nuclear.

Claudin 12 Polyclonal antibody specifically detects Claudin 12 in Human samples. It is validated for Western Blot

Specifications

Antigen Claudin 12
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene CLDN12
Gene Accession No. P56749
Gene Alias claudin 12; claudin-12; Cldn12; RGD1561053
Host Species Rabbit
Immunogen A synthetic peptide from C-terminus of human claudin 12 protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9069
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.