Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claudin-15 Antibody (540315) [Biotin], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB10344B
Description
Claudin-15 Monoclonal antibody specifically detects Claudin-15 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
Claudin-15 | |
Monoclonal | |
Biotin | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
24146 | |
Store at 4°C in the dark. | |
IgG2a |
Flow Cytometry, CyTOF | |
540315 | |
Flow Cytometry, CyTOF-ready | |
claudin 15, claudin-15, FLJ42715, MGC19536 | |
NS0 mouse myeloma cell line transfected with human Claudin-15, Accession # P56746 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction