Learn More
Invitrogen™ Claudin 16 Polyclonal Antibody

Description
Claudin 16 Polyclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | Claudin 16 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | CLDN16 |
| Gene Accession No. | Q9Y5I7 |
| Gene Alias | claudin 16; claudin-16; claudin-16; LOW QUALITY PROTEIN: claudin-16; CLDN16; H59D2a protein; HOMG3; hypomagnesemia 3, with hypercalciuria and nephrocalcinosis; paracellin-1; PCLN1; PCLN-1 |
| Gene Symbols | CLDN16 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.