Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claudin-17 Antibody (410509), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4619L
Description
Claudin-17 Monoclonal antibody specifically detects Claudin-17 in Human samples. It is validated for Flow Cytometry, Flow Cytometry (Intracellular Staining), CyTOFSpecifications
Claudin-17 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
26285 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry, Flow Cytometry (Intracellular Staining), CyTOF | |
410509 | |
Flow Cytometry, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
claudin 17, human CLDN17 gene for claudin-17, 10claudin-17, MGC126552, MGC126554 | |
NS0 mouse myeloma cell line transfected with human Claudin-17, Met1-Val224, Accession # P56750 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction