Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claudin-17 Antibody (410509), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4619H
Description
Claudin-17 Monoclonal antibody specifically detects Claudin-17 in Human samples. It is validated for Flow Cytometry, Flow Cytometry, CyTOFSpecifications
| Claudin-17 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 410509 | |
| Flow Cytometry, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| claudin 17, human CLDN17 gene for claudin-17, 10claudin-17, MGC126552, MGC126554 | |
| NS0 mouse myeloma cell line transfected with human Claudin-17, Met1-Val224, Accession # P56750 | |
| RUO | |
| 26285 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction