Learn More
Claudin 17 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA124419
Description
PA1-24419 detects Claudin 17 in Human samples. PA1-24419 has been successfully used in Western Blot procedures. PA1-24419 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human CLDN17. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
CLDN17, clustered with CLDN8 at human chromosome 21q22.11, is a four-transmembrane protein with WWCC motif, defined by W-X(17-22)-W-X(2)-C-X(8-10)-C.
Specifications
| Claudin 17 | |
| Polyclonal | |
| Unconjugated | |
| P56750 | |
| CLDN17 | |
| Synthetic peptide region of human CLDN17. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| CLDN17 | |
| UNQ758, MGC126552, MGC126554 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 26285 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.