Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claudin 17 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983132
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3132 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3132 Supplier Bioss Supplier No. BS13751R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CLDN17, clustered with CLDN8 at human chromosome 21q22.11, is a four-transmembrane protein with WWCC motif, defined by W-X(17-22)-W-X(2)-C-X(8-10)-C.

Specifications

Antigen Claudin 17
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CLDN17
Gene Accession No. P56750
Gene Alias claudin 17; Claudin-17; Cldn17; human CLDN17 gene for claudin-17; LOC100354623; MGC126552; MGC126554; UNQ758; UNQ758/PRO1489
Gene Symbols CLDN17
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Claudin 17.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26285
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.