Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claudin 18.2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5141108

This item is not returnable.
View return policy
Description
Claudin 18.2 Polyclonal antibody specifically detects Claudin 18.2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
Specifications
Claudin 18.2 | |
Polyclonal | |
Unconjugated | |
CLDN18 | |
SFTA5; SFTPJ | |
Synthetic peptide corresponding to N terminal of Human CLDN18. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
P56856, P56857 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
315953, 51208, 56492 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction