Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Claudin 18.2 Recombinant Human Monoclonal Antibody (9C2)
GREENER_CHOICE

Catalog No. PIMA543155
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543155 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA543155 Supplier Invitrogen™ Supplier No. MA543155
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Recombinant Monoclonal Antibody

Antibody tested in Indirect ELISA.

Plays a major role in tight junction-specific obliteration of the intercellular space, through calcium-independent cell-adhesion activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Claudin 18.2
Applications ELISA, Flow Cytometry
Classification Recombinant Monoclonal
Clone 9C2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CLDN18
Gene Accession No. P56856
Gene Alias SFTA5; SFTPJ
Gene Symbols CLDN18
Host Species Human
Immunogen Recombinant Human Claudin-18.2 protein.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51208
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.