Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claudin-18 Antibody (25E3), Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP32665750UL
Description
Claudin-18 Monoclonal antibody specifically detects Claudin-18 in Human samples. It is validated for Flow Cytometry, ELISASpecifications
| Claudin-18 | |
| Monoclonal | |
| Unconjugated | |
| 50% Glycerol, 0.01M PBS, PH 7.4 | |
| Human | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA | |
| 25E3 | |
| Flow Cytometry 1:50-1:200, ELISA | |
| claudin 18, SFTA5, SFTPJ, surfactant associated 5, surfactant associated protein J, surfactant, pulmonary associated protein J | |
| Recombinant Human Claudin-18 protein [UniProt P56856] | |
| 50 μL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 51208 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction