Learn More
Invitrogen™ Claudin 19 Polyclonal Antibody

Description
Immunogen sequence: GDAIITAVGL YEGLWMSCAS QSTGQVQCKL YDSLL Antigen sequence identity: Mouse 100%; Rat 100%.
Specifications
Specifications
| Antigen | Claudin 19 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CLDN19 |
| Gene Accession No. | Q8N6F1 |
| Gene Alias | claudin 19; claudin-19; Cldn19; HOMG5 |
| Gene Symbols | CLDN19 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.