Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Claudin 20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597593
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597593 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597593 Supplier Invitrogen™ Supplier No. PA597593
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Claudin 20 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

The Claudin 20 gene encodes a member of the claudin family. Claudins are integral membrane proteins and components of tight junction strands. Tight junction strands serve as a physical barrier to prevent solutes and water from passing freely through the paracellular space between epithelial or endothelial cell sheets, and also play critical roles in maintaining cell polarity and signal transductions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Claudin 20
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CLDN20
Gene Accession No. P56880
Gene Alias claudin 20; claudin-20; CLDN20; EG621628; testis secretory sperm-binding protein Li 229n
Gene Symbols CLDN20
Host Species Rabbit
Immunogen Recombinant Human Claudin-20 protein (29-81AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 49861
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.