Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Claudin 25 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562804
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562804 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562804 Supplier Invitrogen™ Supplier No. PA562804
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GIWEVCVDRE EVATVCKAFE SFLSL Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 72%.

Plays a major role in tight junction-specific obliteration of the intercellular space, through calcium-independent cell-adhesion activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Claudin 25
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CLDN25
Gene Accession No. C9JDP6
Gene Alias claudin 25; CLDN25; Putative claudin-25
Gene Symbols CLDN25
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Claudin 25. Recombinant protein control fragment (Product #RP-100675).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 644672
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.