Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claudin-3 Antibody (385013), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB46201Z
Description
Claudin-3 Monoclonal antibody specifically detects Claudin-3 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
Claudin-3 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
385013 | |
Flow Cytometry, CyTOF-ready | |
C7orf1, claudin 3, claudin-3, Clostridium perfringens enterotoxin receptor 2, CPE-R 2, CPE-R2, CPETR2CPE-receptor 2, HRVP1, Rat ventral prostate.1 protein homolog, RVP1, ventral prostate.1 protein homolog, ventral prostate.1-like protein | |
HEK293 human embryonic kidney cell line transfected with human Claudin-3, Met1-Val220, Accession # O15551 | |
0.1 mL | |
Cell Biology, Cellular Markers, Extracellular Matrix | |
1365 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction