Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claudin-3 Antibody (OTI1E7), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270445B
Description
Claudin-3 Monoclonal specifically detects Claudin-3 in Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Claudin-3 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
CLDN3 | |
Full length recombinant protein of human Claudin-3 (NP_001297) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Zebrafish | |
IgG2b |
Western Blot, Immunohistochemistry | |
OTI1E7 | |
Western Blot, Immunohistochemistry | |
C7orf1, claudin 3, claudin-3, Clostridium perfringens enterotoxin receptor 2, CPE-R 2, CPE-R2, CPETR2CPE-receptor 2, HRVP1, Rat ventral prostate.1 protein homolog, RVP1, ventral prostate.1 protein homolog, ventral prostate.1-like protein | |
Mouse | |
Affinity Purified | |
Cell Biology, Cytoskeleton Markers, Extracellular Matrix | |
1365.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction