Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Claudin 3 Monoclonal Antibody (CL13130)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA546539
This item is not returnable.
View return policy
Description
Claudin 3 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Claudin 1-8 proteins are a family of transmembrane proteins associated with tight junctions. Tight junctions are specialized regions of cell to cell contact; made up of network of strands to act as a molecular gasket for preventing the leakage of ions, water etc. between cells. They are abundant in luminal epithelial sheets where they maintain epithelial cell polarity. Different tissues exhibit different Claudin composition. Differential expression of this gene has been observed in different types of malignancies, including breast cancer, ovarian cancer, hepatocellular carcinomas, urinary tumors, prostate cancer, lung cancer, head and neck cancers, thyroid carcinomas, etc. Alternatively spliced transcript variants encoding different isoforms have been found.
Specifications
Claudin 3 | |
Monoclonal | |
0.97 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O15551 | |
CLDN3 | |
Synthetic Peptide. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
CL13130 | |
Unconjugated | |
CLDN3 | |
AI182374; C7orf1; claudin 3; claudin-3; CLDN3; clostridium perfringens enterotoxin receptor 2; CPE-R 2; CPE-R2; CPE-receptor 2; CPETR2; hRVP1; integral membrane protein claudin-3; mRVP1; Rat ventral prostate.1 protein; Rat ventral prostate.1 protein homolog; RVP1; tight junction protein; ventral prostate.1 protein; ventral prostate.1 protein homolog; ventral prostate.1-like protein | |
Mouse | |
Protein A | |
RUO | |
1365 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction