Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claudin 3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA516867
Description
PA5-16867 targets Claudin 3 in IHC (P) and WB applications and shows reactivity with Human, mouse, and Rat samples. The PA5-16867 immunogen is a synthetic peptide derived from the C-terminus of mouse Claudin-3.
Claudin 1-8 proteins are a family of transmembrane proteins associated with tight junctions. Tight junctions are specialized regions of cell to cell contact; made up of network of strands to act as a molecular gasket for preventing the leakage of ions, water etc. between cells. They are abundant in luminal epithelial sheets where they maintain epithelial cell polarity. Different tissues exhibit different Claudin composition.
Specifications
Claudin 3 | |
Polyclonal | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.6 | |
O15551, Q9Z0G9, Q63400 | |
CLDN3 | |
A synthetic peptide derived from the C-terminus of mouse Claudin-3 | |
500 μL | |
Primary | |
Chicken, Human, Mouse, Pig, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
CLDN3 | |
CPE-R 2, CPE-receptor 2, Clostridium perfringens enterotoxin receptor 2, claudin-3, ventral prostate.1 protein homolog, ventral prostate.1-like protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
12739, 1365, 65130 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction