Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Claudin 3 Recombinant Human Monoclonal Antibody (12B2)
GREENER_CHOICE

Catalog No. FEMA550083
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550083 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550083 Supplier Invitrogen™ Supplier No. MA550083
Add to Cart
Add to Cart

Human Recombinant Monoclonal Antibody

Claudin 3 Recombinant Monoclonal Antibody for ELISA

Claudin 1-8 proteins are a family of transmembrane proteins associated with tight junctions. Tight junctions are specialized regions of cell to cell contact; made up of network of strands to act as a molecular gasket for preventing the leakage of ions, water etc. between cells. They are abundant in luminal epithelial sheets where they maintain epithelial cell polarity. Different tissues exhibit different Claudin composition. Differential expression of this gene has been observed in different types of malignancies, including breast cancer, ovarian cancer, hepatocellular carcinomas, urinary tumors, prostate cancer, lung cancer, head and neck cancers, thyroid carcinomas, etc. Alternatively spliced transcript variants encoding different isoforms have been found.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Claudin 3
Applications ELISA, Neutralization
Classification Recombinant Monoclonal
Clone 12B2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene CLDN3
Gene Accession No. O15551
Gene Alias AI182374; C7orf1; claudin 3; claudin-3; CLDN3; clostridium perfringens enterotoxin receptor 2; CPE-R 2; CPE-R2; CPE-receptor 2; CPETR2; hRVP1; integral membrane protein claudin-3; mRVP1; Rat ventral prostate.1 protein; Rat ventral prostate.1 protein homolog; RVP1; tight junction protein; ventral prostate.1 protein; ventral prostate.1 protein homolog; ventral prostate.1-like protein
Gene Symbols CLDN3
Host Species Human
Immunogen Recombinant Human CLDN3 protein.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1365
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.