Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Claudin 4 Recombinant Mouse Monoclonal Antibody (10F4)
GREENER_CHOICE

Catalog No. FEMA550208
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550208 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550208 Supplier Invitrogen™ Supplier No. MA550208
Add to Cart
Add to Cart

Mouse Recombinant Monoclonal Antibody

Claudin 4 Recombinant Monoclonal Antibody for Flow, ELISA

Claudin-4 belongs to the 20-member family of claudins, transmembrane cellular adhesion proteins localized at epithelial and endothelial tight junctions. Claudins demonstrate distinct, tissue-specific expression patterns, and are essential components of tight junction charge-specific channels that regulate paracellular ion flux. Claudin-4 is a component of tight junction strands and may play a role in internal organ development and function during pre- and postnatal life. The overexpression of Claudin-4 has been shown to decrease paracellular electrical conductance due to a selective decrease in Na+ permeability, with no significant decrease in Cl ;- ; permeability. In ovarian carcinoma cells, Claudin-3 and Claudin-4 exhibit upregulation at both mRNA and protein levels. Claudin-4 is also overexpressed in pancreatic cancer tissues and cell lines, and in other epithelial cancers. The Claudin-4 gene is deleted in Williams-Beuren syndrome, a neurodevelopmental disorder affecting multiple systems.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Claudin 4
Applications ELISA, Flow Cytometry
Classification Recombinant Monoclonal
Clone 10F4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene Cldn4
Gene Accession No. O14493
Gene Alias Cep-r; claudin 4; CLAUDIN4; claudin-4; CLDN4; Clostridium perfringens enterotoxin receptor; Clostridium perfringens enterotoxin receptor 1; CPER; CPE-R; CPE-receptor; Cpetr; CPETR1; h CPE-receptor; hCPE-R; WBSCR8; Williams-Beuren syndrome chromosomal region 8 protein
Gene Symbols Cldn4
Host Species Mouse
Immunogen A synthesized peptide derived from Human CLDN4.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1364
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.