Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claudin-5 Antibody (OTI1G4), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271329JF549
Description
Claudin-5 Monoclonal specifically detects Claudin-5 in Human, Mouse, Rat, Porcine samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
Claudin-5 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CLDN5 | |
Full length recombinant protein of human Claudin-5 (NP_003268) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Pig | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI1G4 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
AWALTransmembrane protein deleted in VCFS, BEC1, claudin 5, claudin-5, CPETRL1, TMVCFTMDVCF, transmembrane protein deleted in velocardiofacial syndrome | |
Mouse | |
Affinity Purified | |
Cellular Markers | |
7122.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction