Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claudin-5 Antibody (OTI1G4), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271329MFV500
Description
Claudin-5 Monoclonal antibody specifically detects Claudin-5 in Human, Mouse, Rat, Porcine samples. It is validated for Flow CytometrySpecifications
Claudin-5 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig | |
Purified |
Flow Cytometry | |
OTI1G4 | |
Flow Cytometry | |
AWALTransmembrane protein deleted in VCFS, BEC1, claudin 5, claudin-5, CPETRL1, TMVCFTMDVCF, transmembrane protein deleted in velocardiofacial syndrome | |
Full length human recombinant protein of human CLDN5(NP_003268) produced in HEK293T cell. | |
0.1 mL | |
Cellular Markers | |
7122 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction