Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claudin-5 Antibody (OTI1G4), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP202259
Description
Claudin-5 Monoclonal specifically detects Claudin-5 in Human, Mouse, Rat, Porcine samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
Claudin-5 | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunocytochemistry | |
CLDN5 | |
Full length human recombinant protein of human CLDN5(NP_003268) produced in HEK293T cell. | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI1G4 | |
Unconjugated | |
AWALTransmembrane protein deleted in VCFS, BEC1, claudin 5, claudin-5, CPETRL1, TMVCFTMDVCF, transmembrane protein deleted in velocardiofacial syndrome | |
Mouse | |
23 kDa | |
0.1 mL | |
Cellular Markers | |
7122 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction