Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Claudin 6/9 Recombinant Human Monoclonal Antibody (9H8)
Human Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA550081
Description
Claudin 6/9 Recombinant Monoclonal Antibody for Western Blot, Flow, ELISA
Plays a major role in tight junction-specific obliteration of the intercellular space.
Specifications
Claudin 6/9 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
P56747 | |
CLDN6 | |
Recombinant Human CLDN6/9 protein (aa 1-217). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Western Blot | |
9H8 | |
Unconjugated | |
CLDN6 | |
Claudin6; Claudin-6; CLD6_HUMAN; CLDN 6; CLDN6; OTTHUMP00000159248; Skullin; Skullin 2; UNQ757/PRO1488 | |
Human | |
Affinity chromatography | |
RUO | |
9074 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction