Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claudin-6 Antibody (342927), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB3656D
Description
Claudin-6 Monoclonal antibody specifically detects Claudin-6 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
Claudin-6 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry, CyTOF | |
342927 | |
Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
claudin 6, claudin-6, skullin | |
NS0 mouse myeloma cell line transfected with human Claudin-6, Met1-Val220, Accession # P56747 | |
0.1 mL | |
Stem Cell Markers | |
9074 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction