Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claudin-6 Antibody (OTI2H1), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271416AF700
Description
Claudin-6 Monoclonal specifically detects Claudin-6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Claudin-6 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CLDN6 | |
Full length recombinant protein of human Claudin-6 (NP_067018) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI2H1 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
claudin 6, claudin-6, skullin | |
Mouse | |
Affinity Purified | |
Stem Cell Markers | |
9074.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction