Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claudin-6 Antibody (OTI2H1) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271416
Description
Claudin-6 Monoclonal specifically detects Claudin-6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Claudin-6 | |
Monoclonal | |
LYOPH | |
Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:100 | |
claudin 6, claudin-6, skullin | |
Mouse | |
23.1 kDa | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI2H1 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
CLDN6 | |
Full length human recombinant protein of human CLDN6(NP_067018) produced in HEK293T cell. | |
Affinity Purified | |
Stem Cell Markers | |
9074 | |
Human, Mouse, Rat | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction