Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Claudin 6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120728
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120728 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120728 Supplier Invitrogen™ Supplier No. PA5120728
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: OVCAR3, Rat brain. Immunogen sequence: SGIVFVISGV LTLIPVCWTA HAIIRDFYNP LVAEAQKREL GASLYLGWAA SGLLLLGGGL LCCTCPSGGS QGPSHYMARY STSAPAISRG PSEYPTKNYV.

Tight junctions represent one mode of cell-to-cell adhesion in epithelial or endothelial cell sheets, forming continuous seals around cells and serving as a physical barrier to prevent solutes and water from passing freely through the paracellular space. These junctions are comprised of sets of continuous networking strands in the outwardly facing cytoplasmic leaflet, with complementary grooves in the inwardly facing extracytoplasmic leaflet. This gene encodes a component of tight junction strands, which is a member of the claudin family. The protein is an integral membrane protein and is one of the entry cofactors for hepatitis C virus. The gene methylation may be involved in esophageal tumorigenesis. This gene is adjacent to another family member CLDN9 on chromosome 16.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Claudin 6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.76 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene CLDN6
Gene Accession No. P56747, Q9Z262
Gene Alias AL024037; claudin 6; Claudin6; claudin-6; CLD6; CLDN6; Skullin; tight junction molecule claudin 6; UNQ757/PRO1488
Gene Symbols CLDN6
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 287098, 54419, 9074
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.