Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claudin-7 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318508V
Description
Claudin-7 Polyclonal antibody specifically detects Claudin-7 in Human samples. It is validated for Western BlotSpecifications
Claudin-7 | |
Polyclonal | |
Western Blot | |
CEPTRL2, claudin 7, claudin-1, claudin-7, CLDN-7, clostridium perfringens enterotoxin receptor-like 2, CPETRL2 | |
Recognizes region between residues Between between 161 and 211.. | |
0.1 mL | |
Breast Cancer, Cancer | |
1366 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction