Learn More
Invitrogen™ Claudin 7 Polyclonal Antibody
Rabbit Polyclonal Antibody
$516.00 - $743.00
Specifications
Antigen | Claudin 7 |
---|---|
Concentration | 0.12 mg/mL |
Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
01-673-305
|
Invitrogen™
PA137474 |
1 mL |
Each for $743.00
|
|
|||||
PIPA532356
|
Invitrogen™
PA532356 |
500 μL |
Each for $516.00
|
|
|||||
Description
This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is colon carcinoma.
This gene encodes a member of the claudin family. Claudins are integral membrane proteins and components of tight junction strands. Tight junction strands serve as a physical barrier to prevent solutes and water from passing freely through the paracellular space between epithelial or endothelial cell sheets, and also play critical roles in maintaining cell polarity and signal transductions. Differential expression of this gene has been observed in different types of malignancies, including breast cancer, ovarian cancer, hepatocellular carcinomas, urinary tumors, prostate cancer, lung cancer, head and neck cancers, thyroid carcinomas, etc. Alternatively spliced transcript variants encoding different isoforms have been found.Specifications
Claudin 7 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Polyclonal | |
Liquid | |
IgG | |
Human, Mouse, Rat | |
CLDN7 | |
CEPTRL2; claudin 7; claudin-1; Claudin-7; Cldn7; CLDN-7; clostridium perfringens enterotoxin receptor-like 2; CPETRL2; Hs.84359 | |
CLDN7 | |
Primary | |
Antigen Affinity Chromatography |
0.12 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.6 | |
O95471, Q9Z1L1, Q9Z261 | |
1366, 53624, 65132 | |
Synthetic peptide from C-terminus of human claudin 7 protein. | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.