Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claudin-8 Antibody (474210), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB5275F
Description
Claudin-8 Monoclonal antibody specifically detects Claudin-8 in Human samples. It is validated for Flow Cytometry, Flow Cytometry (Intracellular Staining), CyTOFSpecifications
Claudin-8 | |
Monoclonal | |
FITC | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
9073 | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry, Flow Cytometry (Intracellular Staining), CyTOF | |
474210 | |
Flow Cytometry, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
claudin 8, human CLDN8 gene for claudin-810claudin-8 | |
HEK293 human embryonic kidney cell line transfected with human Claudin-8, Met1-Val225 (Ser151Pro), Accession # P56748 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction