Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Claudin 9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104427
Description
Antibody detects endogenous levels of total Claudin 9.
CLDN9 is a member of claudin family. It is a four-transmembrane protein with WWCC motif, defined by W-X(17-22)-W-X(2)-C-X(8-10)-C.
Specifications
Claudin 9 | |
Polyclonal | |
Unconjugated | |
Cldn9 | |
claudin 9; claudin-9; CLDN9; nmf329 | |
Rabbit | |
Affinity chromatography | |
RUO | |
287099, 56863, 9080 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O95484, Q9Z0S7 | |
Cldn9 | |
A synthesized peptide derived from human CLDN9(Accession O95484), corresponding to amino acid residues G179-V217. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction