Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Clavesin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA535378
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535378 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535378 Supplier Invitrogen™ Supplier No. PA535378
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with human and mouse based on 100% sequence homology. This antibody contains enough material to conduct 10 mini-Western blots.

Required for normal morphology of late endosomes and/or lysosomes in neurons. Binds phosphatidylinositol 3,5-bisphosphate (PtdIns(3,5)P2).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Clavesin
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and no preservative
Gene Clvs1
Gene Accession No. A6JFQ6, A6JUQ6
Gene Alias 4933402J24Rik; A330019N05Rik; bA160A10.4; C6orf212; C6orf213; Cellular retinaldehyde-binding protein-like; clathrin vesicle-associated Sec14 protein 1; clathrin vesicle-associated Sec14 protein 2; clavesin 1; clavesin 2; Clavesin-1; clavesin-1 {ECO:0000303; Clavesin-2; clavesin-2 {ECO:0000303; Clvl1; Clvs1; clvs1 {ECO:0000303; CLVS2; clvs2 {ECO:0000303; CRALBPL; PubMed:19651769}; retinaldehyde binding protein 1-like 1; retinaldehyde binding protein 1-like 2; retinaldehyde-binding protein 1-like 1; Retinaldehyde-binding protein 1-like 2; Retinaldehyde-binding protein 1-like protein 1; retinaldehyde-binding protein 1-like protein 2; RGD1306801; RGD1564200; Rlbp1l1; Rlbp1l2
Gene Symbols Clvs1, Clvs2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminal region of rat CLVS1/2 conjugated to KLH.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361459, 366311
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.