Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLC-5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576761
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576761 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576761 Supplier Invitrogen™ Supplier No. PA576761
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Proton-coupled chloride transporter. Functions as antiport system and exchanges chloride ions against protons. Important for normal acidification of the endosome lumen. May play an important role in renal tubular function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLC-5
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene CLCN5
Gene Accession No. P51795, Q9WVD4
Gene Alias 5430408K11Rik; chloride channel 5; chloride channel 5 (nephrolithiasis 2, X-linked, Dent disease); chloride channel ClC5; chloride channel Clc-5; chloride channel CLCN5; chloride channel protein 5; chloride channel, voltage-sensitive 5; chloride transporter ClC-5; chloride voltage-gated channel 5; Clc5; clC-5; CLCK2; CLCN3; CLCN4; Clcn5; D930009B12Rik; DENTS; DXImx42e; H(+)/Cl(-) exchange transporter 5; H(+)/Cl(-) exchange transporter 5-like; hCIC-K2; hClC-K2; NPHL1; NPHL2; Sfc13; T25545; voltage-gated chloride ion channel CLCN5; XLRH; XRN
Gene Symbols CLCN5
Host Species Rabbit
Immunogen Recombinant full length Human CLCN5.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1184, 12728
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.