Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLC-5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102830
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102830 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102830 Supplier Invitrogen™ Supplier No. PA5102830
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CLCN5.

Proton-coupled chloride transporter. Functions as antiport system and exchanges chloride ions against protons. Important for normal acidification of the endosome lumen. May play an important role in renal tubular function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLC-5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CLCN5
Gene Accession No. P51795, P51796, Q9WVD4
Gene Alias 5430408K11Rik; chloride channel 5; chloride channel 5 (nephrolithiasis 2, X-linked, Dent disease); chloride channel ClC5; chloride channel Clc-5; chloride channel CLCN5; chloride channel protein 5; chloride channel, voltage-sensitive 5; chloride transporter ClC-5; chloride voltage-gated channel 5; Clc5; clC-5; CLCK2; CLCN3; CLCN4; Clcn5; D930009B12Rik; DENTS; DXImx42e; H(+)/Cl(-) exchange transporter 5; H(+)/Cl(-) exchange transporter 5-like; hCIC-K2; hClC-K2; NPHL1; NPHL2; Sfc13; T25545; voltage-gated chloride ion channel CLCN5; XLRH; XRN
Gene Symbols CLCN5
Host Species Rabbit
Immunogen A synthesized peptide derived from human CLCN5(Accession P51795), corresponding to amino acid residues L384-T434.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1184, 12728, 25749
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.