Learn More
CLCA1 Mouse anti-Human, Clone: OTI8E7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809864

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the calcium sensitive chloride conductance protein family. To date, all members of this gene family map to the same region on chromosome 1p31-p22 and share a high degree of homology in size, sequence, and predicted structure, but differ significantly in their tissue distributions. The encoded protein is expressed as a precursor protein that is processed into two cell-surface-associated subunits, although the site at which the precursor is cleaved has not been precisely determined. The encoded protein may be involved in mediating calcium-activated chloride conductance in the intestine.Specifications
CLCA1 | |
Monoclonal | |
Unconjugated | |
CLCA1 | |
CACC, CACC1, CLCRG1, CaCC-1, GOB5, hCLCA1, hCaCC-1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
1179 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI8E7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
A8K7I4 | |
CLCA1 | |
Human recombit protein fragment corresponding to amino acids 344-412 of human CLCA1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.