Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLCA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562682
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562682 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562682 Supplier Invitrogen™ Supplier No. PA562682
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VGMVTFDSAA HVQSELIQIN SGSDRDTLAK RLPAAASGGT SICSGLRSAF TVIRKKYPTD GSEIVLLTD Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 80%.

This gene encodes a member of the calcium sensitive chloride conductance protein family. To date, all members of this gene family map to the same region on chromosome 1p31-p22 and share a high degree of homology in size, sequence, and predicted structure, but differ significantly in their tissue distributions. The encoded protein is expressed as a precursor protein that is processed into two cell-surface-associated subunits, although the site at which the precursor is cleaved has not been precisely determined. The encoded protein may be involved in mediating calcium-activated chloride conductance in the intestine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLCA1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CLCA1
Gene Accession No. A8K7I4
Gene Alias CACC; CACC1; CaCC-1; Calcium-activated chloride channel family member 1; calcium-activated chloride channel family member 3; Calcium-activated chloride channel protein 1; calcium-activated chloride channel regulator 1; calcium-dependent chloride channel-1; chloride channel accessory 1; chloride channel calcium activated 1; chloride channel calcium activated 2; chloride channel calcium activated 3; chloride channel regulator 1; chloride channel, calcium activated, family member 1; CLCA family member 1, chloride channel regulator; Clca1; Clca2; Clca3; CLCRG1; Gob5; gob-5; hCaCC-1; hCLCA1; mCLCA3; Protein gob-5
Gene Symbols CLCA1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CLCA1. Recombinant protein control fragment (Product #RP-101092).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1179
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.