Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLCA3P Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA549089

Catalog No. PA549089


Only null left
Add to Cart

Description

Description

CLCA3P Polyclonal Antibody for Western Blot

This gene is a transcribed pseudogene belonging to the calcium sensitive chloride conductance protein family. To date, all members of this gene family map to the same site on chromosome 1p31-p22 and share high degrees of homology in size, sequence and predicted structure, but differ significantly in their tissue distributions. This gene contains several nonsense codons compared to other family members that render the transcript a candidate for nonsense-mediated mRNA decay (NMD). Therefore, this gene is unlikely to be protein-coding. [provided by RefSeq, Jan 2009].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CLCA3P
Polyclonal
PBS with 0.09% sodium azide; pH 7.4
Q9Y6N3
CLCA3P
KLH conjugated synthetic peptide between 70-98 amino acids from the Central region of human CLCA3P.
400 μL
Primary
Human
Antibody
IgG
Western Blot
Unconjugated
CLCA3P
chloride channel accessory 3, pseudogene; CLCA3; CLCA3P; hCLCA3
Rabbit
Antigen Affinity Chromatography, Protein A
RUO
9629
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.