Learn More
Invitrogen™ CLCA4 Polyclonal Antibody

Description
The antibody detects endogenous levels of total CLCA4 protein.
Specifications
Specifications
| Antigen | CLCA4 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | CLCA4 |
| Gene Accession No. | Q14CN2 |
| Gene Alias | CaCC; CaCC2; CaCC-2; Calcium-activated chloride channel family member 4; calcium-activated chloride channel protein 2; Calcium-activated chloride channel regulator 4; Calcium-activated chloride channel regulator 4, 110 kDa form; Calcium-activated chloride channel regulator 4, 30 kDa form; Chloride channel accessory 4; chloride channel regulator 4; chloride channel, calcium activated, family member 4; CLCA4; hCaCC-2; hCLCA4; UNQ562/PRO1124 |
| Gene Symbols | CLCA4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.