Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLCA4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139869
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139869 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5139869 Supplier Invitrogen™ Supplier No. PA5139869
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: NEDQPFYRAK SKKIEATRCS AGISGRNRVY KCQGGSCLSR ACRIDSTTKL YGKDCQFFPD KVQT Highest antigen sequence indentity to the following orthologs: Mouse - 80%, Rat - 75%.

The protein encoded by this gene belongs to the calcium sensitive chloride conductance protein family. To date, all members of this gene family map to the same site on chromosome 1p31-p22 and share high degrees of homology in size, sequence and predicted structure, but differ significantly in their tissue distributions. Alternative splicing results in multiple transcript variants, only one of which is thought to be protein coding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLCA4
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CLCA4
Gene Accession No. Q14CN2
Gene Alias CaCC; CaCC2; CaCC-2; Calcium-activated chloride channel family member 4; calcium-activated chloride channel protein 2; Calcium-activated chloride channel regulator 4; Calcium-activated chloride channel regulator 4, 110 kDa form; Calcium-activated chloride channel regulator 4, 30 kDa form; Chloride channel accessory 4; chloride channel regulator 4; chloride channel, calcium activated, family member 4; CLCA4; hCaCC-2; hCLCA4; UNQ562/PRO1124
Gene Symbols CLCA4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CLCA4. Recombinant protein control fragment (Product #RP-109070).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22802
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.