Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLCC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552651
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552651 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552651 Supplier Invitrogen™ Supplier No. PA552651
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PPQALRPRDR RRQEEIDYRP DGGAGDADFH YRGQMGPTEQ GPYAKTYEGR REILRERDVD LRFQTGNKSP EVLRAFDVPD AEAREHPTVV PSHKSPVLDT KPKE Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 64%.

Neurofibromin is a member of the GTPase-activating (GAP) class of proteins. These proteins enhance the GTPase activity of signaling G-Proteins like Ras. The NF1 gene is also classified as a tumor suppressor gene, and many studies have shown that mutations at the NF1 locus produce neurofibromas, rhabdomyosarcomas, leimyosarcomas, mental retardation and memory deficiency. Neurofibromin has also been associated with the intermediated filaments of the cytoskeleton. NF1 is expressed at its highest levels during periods of desmosome formation and mutations of NF1 produce variable cell sizes and morphologies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLCC1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CLCC1
Gene Accession No. Q96S66
Gene Alias chloride channel CLIC like 1; chloride channel CLIC-like 1; chloride channel CLIC-like protein 1; CLCC1; KIAA0761; MCLC; Mid-1-related chloride channel 1; mid-1-related chloride channel protein 1
Gene Symbols CLCC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CLCC1. Recombinant protein control fragment (Product #RP-90166).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23155
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.