Learn More
Invitrogen™ CLCC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier:  Invitrogen™ PA598800
Catalog No. PIPA598800
Only null left
Add to Cart
Description
Description
CLCC1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP, ELISA
Neurofibromin is a member of the GTPase-activating (GAP) class of proteins. These proteins enhance the GTPase activity of signaling G-Proteins like Ras. The NF1 gene is also classified as a tumor suppressor gene, and many studies have shown that mutations at the NF1 locus produce neurofibromas, rhabdomyosarcomas, leimyosarcomas, mental retardation and memory deficiency. Neurofibromin has also been associated with the intermediated filaments of the cytoskeleton. NF1 is expressed at its highest levels during periods of desmosome formation and mutations of NF1 produce variable cell sizes and morphologies.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CLCC1
Polyclonal
Unconjugated
CLCC1
chloride channel CLIC like 1; chloride channel CLIC-like 1; chloride channel CLIC-like protein 1; CLCC1; KIAA0761; MCLC; Mid-1-related chloride channel 1; mid-1-related chloride channel protein 1
Rabbit
Protein G
RUO
23155
-20°C or -80°C if preferred
Liquid
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
4.59 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Q96S66
CLCC1
Recombinant Human Chloride channel CLIC-like protein 1 protein (446-551AA).
100 μg
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.