Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLCC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120569
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120569 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120569 Supplier Invitrogen™ Supplier No. PA5120569
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: U-87MG, 22Rv1, Mouse spleen. Immunogen sequence: GAGKSVHVLR HIGGPESEPP QALRPRDRRR QEEIDYRPDG GAGDADFHYR GQMGPTEQGP YAKTYEGRRE ILRERDVDLR FQTGNKSPEV LRAFDVPDAE AREHPTVVPS HKSPVLDTKP KETGGILGEG TPKESSTESS QSAKPVSGQD TSGNTEGSPA AEKAQLKSEA AGSPDQGSTY SPARGVAGPR GQDPVSSPCG.

Neurofibromin is a member of the GTPase-activating (GAP) class of proteins. These proteins enhance the GTPase activity of signaling G-Proteins like Ras. The NF1 gene is also classified as a tumor suppressor gene, and many studies have shown that mutations at the NF1 locus produce neurofibromas, rhabdomyosarcomas, leimyosarcomas, mental retardation and memory deficiency. Neurofibromin has also been associated with the intermediated filaments of the cytoskeleton. NF1 is expressed at its highest levels during periods of desmosome formation and mutations of NF1 produce variable cell sizes and morphologies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLCC1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CLCC1
Gene Accession No. Q96S66, Q99LI2
Gene Alias chloride channel CLIC like 1; chloride channel CLIC-like 1; chloride channel CLIC-like protein 1; CLCC1; KIAA0761; MCLC; Mid-1-related chloride channel 1; mid-1-related chloride channel protein 1
Gene Symbols CLCC1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 302-501 of human CLCC1 (NP_055942.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 229725, 23155
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.