Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CLCN2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553189
Description
Immunogen sequence: GVDHAYVTSI GRLIGIVTLK ELRKAIEGSV TAQGVKVRPP LASFRDSATS SSDTETTEVH ALWGPHSRHG Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
The transmembrane protein encoded by this gene is a voltage-gated chloride channel that maintains chloride ion homeostasis in various cells. Defects in this gene may be a cause of certain epilepsies. Four transcript variants encoding different isoforms have been found for this gene.
Specifications
| CLCN2 | |
| Polyclonal | |
| Unconjugated | |
| CLCN2 | |
| AL118368; chloride channel 2; chloride channel protein 2; chloride channel, voltage-sensitive 2; chloride voltage-gated channel 2; CIC 2; CIC2; CIC-2; Clc2; ClC-2; CLCN2; ECA2; ECA3; EG13; EGI11; EGI3; EGMA; EJM6; EJM8; LKPAT; nmf240 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1181 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P51788 | |
| CLCN2 | |
| Recombinant protein corresponding to Human CLCN2. Recombinant protein control fragment (Product #RP-90936). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction