Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLCN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596510
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596510 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596510 Supplier Invitrogen™ Supplier No. PA596510
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSLYDSIIRI KKLPYLPELG WGRHQQYRVR VEDIMVRDVP HVALSCTFRD LRLALHRTKG RMLALVESPE SMILLGSIER SQVVALLGAQ LSPARRRQHM QERRATQTSP LSDQEGPPTP EASVCFQVNT EDSAFPAARG ETHKPLKPAL KRGPSVTRNL GESPTGSAES AGIALRSLFC GSPPPEAASE KLESCEKRKL KRVRISLASD ADLEGEMSPE E Positive Samples: Mouse brain, Mouse testis; Cellular Location: Cell membrane, Multi-pass membrane protein.

The transmembrane protein encoded by this gene is a voltage-gated chloride channel that maintains chloride ion homeostasis in various cells. Defects in this gene may be a cause of certain epilepsies. Four transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLCN2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.11 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CLCN2
Gene Accession No. P51788, Q9R0A1
Gene Alias AL118368; chloride channel 2; chloride channel protein 2; chloride channel, voltage-sensitive 2; chloride voltage-gated channel 2; CIC 2; CIC2; CIC-2; Clc2; ClC-2; CLCN2; ECA2; ECA3; EG13; EGI11; EGI3; EGMA; EJM6; EJM8; LKPAT; nmf240
Gene Symbols CLCN2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 550-770 of human CLCN2 (NP_0011645601).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1181, 12724
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.