Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLCN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119888
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119888 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119888 Supplier Invitrogen™ Supplier No. PA5119888
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: HepG2 cell lysate, 293 cell lysate, human kidney tissue, human stomach tissue, rat colon tissue. Subcellular Location: Cell membrane.

The transmembrane protein encoded by this gene is a voltage-gated chloride channel that maintains chloride ion homeostasis in various cells. Defects in this gene may be a cause of certain epilepsies. Four transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLCN2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene CLCN2
Gene Accession No. P35525, P51788
Gene Alias AL118368; chloride channel 2; chloride channel protein 2; chloride channel, voltage-sensitive 2; chloride voltage-gated channel 2; CIC 2; CIC2; CIC-2; Clc2; ClC-2; CLCN2; ECA2; ECA3; EG13; EGI11; EGI3; EGMA; EJM6; EJM8; LKPAT; nmf240
Gene Symbols CLCN2
Host Species Rabbit
Immunogen Recombinant protein within human ClC2 aa 631-830.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1181, 29232
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.