Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLCN3 Antibody (S258-5), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259379AF488
Description
CLCN3 Monoclonal specifically detects CLCN3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
CLCN3 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CLCN3 | |
Synthetic peptide amino acids 98-115 (cytoplasmic N-terminus) of rat Clcn3 | |
100 μL | |
1182 | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
S258-5 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
chloride channel 3, Chloride channel protein 3, Chloride transporter ClC-3, ClC-3, CLC3, H(+)/Cl(-) exchange transporter 3 | |
Mouse | |
Protein G purified | |
Primary | |
Detects 90kDa. Does not cross-react with Clcn4 or Clcn5 (based on KO validation results). | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction