Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLCN3 Monoclonal Antibody (N258/5)
GREENER_CHOICE

Catalog No. PIMA527709
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527709 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527709 Supplier Invitrogen™ Supplier No. MA527709
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

1 μg/mL of MA5-27709 was sufficient for detection of Clcn3 in 20 μg of rat brain membrane lysate and assayed by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 90kDa. Does not cross-react with Clcn4 or Clcn5 (based on KO validation results). This antibody was formerly sold as clone S258-5.

Mediates the exchange of chloride ions against protons. Functions as antiporter and contributes to the acidification of the endosome and synaptic vesicle lumen, and may thereby affect vesicle trafficking and exocytosis. May play an important role in neuronal cell function through regulation of membrane excitability by protein kinase C. It could help neuronal cells to establish short-term memory.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLCN3
Applications Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone N258/5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Gene CLCN3
Gene Accession No. P51790, P51791, P51792
Gene Alias chloride channel 3; chloride channel protein 3; chloride channel, voltage-sensitive 3; chloride transporter ClC-3; chloride voltage-gated channel 3; CIC3; Clc3; clC-3; CLCN 3; Clcn3; DKFZp564I0463; H(+)/Cl(-) exchange transporter 3; protein kinase C-regulated chloride channel
Gene Symbols CLCN3
Host Species Mouse
Immunogen Synthetic peptide amino acids 98-115 (cytoplasmic N-terminus) of rat Clcn3.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1182, 12725, 84360
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.