Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clcn3 Mouse anti-Human, Rat, FITC, Clone:S258-5, Abnova™

Catalog No. 89333385 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-385 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-385 Supplier Abnova Corporation Supplier No. MAB16803
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of rat Clcn3.

Specifications

Antigen Clcn3
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone S258-5
Conjugate FITC
Description chloride channel 3
Dilution Immunocytochemistry (1:100)
Immunofluorescence (1:100)
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1000)
Western Blot (1:1000)
The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Clcn3
Gene Alias ClC-3
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 98-115 at N-terminus of rat Clcn3.
Quantity 100 μg
Gene ID (Entrez) 84360
Target Species Human, Rat
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.